Anti-CHEK2 Rabbit Monoclonal Antibody [clone: EPR4325]

Lieferant: Abcam
ABCAAB109413-100EA 976 EUR
ABCAAB109413-100
Anti-CHEK2 Rabbit Monoclonal Antibody [clone: EPR4325]
Antikörper
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: monoclonal
Clone: EPR4325
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR