Anti-ICAM3 Rabbit Monoclonal Antibody [clone: EPR3994]
Lieferant: Abcam
ABCAAB109405-100EA 785 EUR
ABCAAB109405-100
Anti-ICAM3 Rabbit Monoclonal Antibody [clone: EPR3994]
Antikörper
Type: Primary
Antigen: ICAM3 (intercellular adhesion molecule 3)
Clonality: monoclonal
Clone: EPR3994
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen