Anti-MAP3K7 Rabbit Monoclonal Antibody [clone: EPR2863]

Lieferant: Abcam
ABCAAB109404-100EA 847 EUR
ABCAAB109404-100
Anti-MAP3K7 Rabbit Monoclonal Antibody [clone: EPR2863]
Antikörper
Type: Primary
Antigen: MAP3K7 (mitogen-activated protein kinase kinase kinase 7)
Clonality: monoclonal
Clone: EPR2863
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR