Anti-RB1 Rabbit Monoclonal Antibody [clone: EPR224]
Lieferant: Abcam
ABCAAB109399-100EA 776 EUR
ABCAAB109399-100
Anti-RB1 Rabbit Monoclonal Antibody [clone: EPR224]
Antikörper
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: monoclonal
Clone: EPR224
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen