Anti-MAPT Rabbit Monoclonal Antibody [clone: EPR2731]

Lieferant: Abcam
ABCAAB109390-100EA 985 EUR
ABCAAB109390-100
Anti-MAPT Rabbit Monoclonal Antibody [clone: EPR2731]
Antikörper
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: EPR2731
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR