Anti-Proteasome 20S alpha 6 Rabbit Monoclonal Antibody [clone: EPR5835]

Lieferant: Abcam
ABCAAB109377-100EA 895 EUR
ABCAAB109377-100
Anti-Proteasome 20S alpha 6 Rabbit Monoclonal Antibody [clone: EPR5835]
Antikörper
Type: Primary
Antigen: Proteasome 20S alpha 6
Clonality: monoclonal
Clone: EPR5835
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR