Anti-FGFR2 Rabbit Monoclonal Antibody [clone: EPR5180]

Lieferant: Abcam
ABCAAB109372-100EA 920 EUR
ABCAAB109372-100
Anti-FGFR2 Rabbit Monoclonal Antibody [clone: EPR5180]
Antikörper
Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: monoclonal
Clone: EPR5180
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR