Anti-TNFRSF12A Rabbit Monoclonal Antibody [clone: EPR3179]

Lieferant: Abcam
ABCAAB109365-100EA 860 EUR
ABCAAB109365-100
Anti-TNFRSF12A Rabbit Monoclonal Antibody [clone: EPR3179]
Antikörper
Type: Primary
Antigen: TNFRSF12A (tumor necrosis factor receptor superfamily, member 12A)
Clonality: monoclonal
Clone: EPR3179
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR