Anti-TRPM8 Rabbit Monoclonal Antibody [clone: EPR4196]

Lieferant: Abcam
ABCAAB109308-100EA 799 EUR
ABCAAB109308-100
Anti-TRPM8 Rabbit Monoclonal Antibody [clone: EPR4196]
Antikörper
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: monoclonal
Clone: EPR4196
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR