Anti-UFM1 Rabbit Monoclonal Antibody [clone: EPR4264]

Lieferant: Abcam
ABCAAB109305-100EA 934 EUR
ABCAAB109305-100
Anti-UFM1 Rabbit Monoclonal Antibody [clone: EPR4264]
Antikörper
Type: Primary
Antigen: UFM1 (ubiquitin-fold modifier 1)
Clonality: monoclonal
Clone: EPR4264
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR