Anti-TBK1 Rabbit Monoclonal Antibody [clone: EPR2867]

Lieferant: Abcam
ABCAAB109272-100EA 888 EUR
ABCAAB109272-100
Anti-TBK1 Rabbit Monoclonal Antibody [clone: EPR2867]
Antikörper
Type: Primary
Antigen: TBK1 (TANK-binding kinase 1)
Clonality: monoclonal
Clone: EPR2867
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR