Anti-PCYT1A Rabbit Monoclonal Antibody [clone: EPR3940]

Lieferant: Abcam
ABCAAB109263-100EA 785 EUR
ABCAAB109263-100
Anti-PCYT1A Rabbit Monoclonal Antibody [clone: EPR3940]
Antikörper
Type: Primary
Antigen: PCYT1A (phosphate cytidylyltransferase 1, choline, alpha)
Clonality: monoclonal
Clone: EPR3940
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR