Anti-LCP2 Rabbit Monoclonal Antibody [clone: EPR2549]

Lieferant: Abcam
ABCAAB109254-100EA 862 EUR
ABCAAB109254-100
Anti-LCP2 Rabbit Monoclonal Antibody [clone: EPR2549]
Antikörper
Type: Primary
Antigen: LCP2 (lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa))
Clonality: monoclonal
Clone: EPR2549
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR