Anti-HSP90AA2 Rabbit Monoclonal Antibody [clone: EPR3953]

Lieferant: Abcam
ABCAAB109248-100EA 906 EUR
ABCAAB109248-100
Anti-HSP90AA2 Rabbit Monoclonal Antibody [clone: EPR3953]
Antikörper
Type: Primary
Antigen: HSP90AA2 (heat shock protein 90kDa alpha (cytosolic), class A member 2)
Clonality: monoclonal
Clone: EPR3953
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR