Anti-SMC1 Rabbit Monoclonal Antibody [clone: EPR2879]
Lieferant: Abcam
ABCAAB109238-100EA 854 EUR
ABCAAB109238-100
Anti-SMC1 Rabbit Monoclonal Antibody [clone: EPR2879]
Antikörper
Type: Primary
Antigen: SMC1
Clonality: monoclonal
Clone: EPR2879
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen