Anti-NEUROG2 Rabbit Monoclonal Antibody [clone: EPR4177]
Lieferant: Abcam
ABCAAB109236-100EA 859 EUR
ABCAAB109236-100
Anti-NEUROG2 Rabbit Monoclonal Antibody [clone: EPR4177]
Antikörper
Type: Primary
Antigen: NEUROG2 (neurogenin 2)
Clonality: monoclonal
Clone: EPR4177
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen