Anti-TYRO3 Rabbit Monoclonal Antibody [clone: EPR4308]

Lieferant: Abcam
ABCAAB109231-100EA 897 EUR
ABCAAB109231-100
Anti-TYRO3 Rabbit Monoclonal Antibody [clone: EPR4308]
Antikörper
Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: monoclonal
Clone: EPR4308
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR