Anti-LTF Rabbit Monoclonal Antibody [clone: EPR4337]

Lieferant: Abcam
ABCAAB109216-100EA 831 EUR
ABCAAB109216-100
Anti-LTF Rabbit Monoclonal Antibody [clone: EPR4337]
Antikörper
Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: monoclonal
Clone: EPR4337
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR