Anti-PDCD6 Rabbit Monoclonal Antibody [clone: EP3259]
Lieferant: Abcam
ABCAAB109181-100EA 854 EUR
ABCAAB109181-100
Anti-PDCD6 Rabbit Monoclonal Antibody [clone: EP3259]
Antikörper
Type: Primary
Antigen: PDCD6 (programmed cell death 6)
Clonality: monoclonal
Clone: EP3259
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen