Anti-TRAM2 Rabbit Monoclonal Antibody [clone: EPR2658]
Lieferant: Abcam
ABCAAB109176-100EA 725 EUR
ABCAAB109176-100
Anti-TRAM2 Rabbit Monoclonal Antibody [clone: EPR2658]
Antikörper
Type: Primary
Antigen: TRAM2 (translocation associated membrane protein 2)
Clonality: monoclonal
Clone: EPR2658
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen