Anti-PTP4A2 Rabbit Monoclonal Antibody [clone: EPR3800]

Lieferant: Abcam
ABCAAB109118-100EA 778 EUR
ABCAAB109118-100
Anti-PTP4A2 Rabbit Monoclonal Antibody [clone: EPR3800]
Antikörper
Type: Primary
Antigen: PTP4A2 (protein tyrosine phosphatase type IVA, member 2)
Clonality: monoclonal
Clone: EPR3800
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR