Anti-MAGE 1 Rabbit Monoclonal Antibody [clone: EPR4276]

Lieferant: Abcam
ABCAAB109108-100EA 882 EUR
ABCAAB109108-100
Anti-MAGE 1 Rabbit Monoclonal Antibody [clone: EPR4276]
Antikörper
Type: Primary
Antigen: MAGE 1
Clonality: monoclonal
Clone: EPR4276
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR