Anti-GSC Rabbit Monoclonal Antibody [clone: EPR4847]

Lieferant: Abcam
ABCAAB109024-100EA 834 EUR
ABCAAB109024-100
Anti-GSC Rabbit Monoclonal Antibody [clone: EPR4847]
Antikörper
Type: Primary
Antigen: GSC (goosecoid homeobox)
Clonality: monoclonal
Clone: EPR4847
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR