Anti-RSF1 Rabbit Monoclonal Antibody [clone: EPR3749]

Lieferant: Abcam
ABCAAB109002-100EA 777 EUR
ABCAAB109002-100
Anti-RSF1 Rabbit Monoclonal Antibody [clone: EPR3749]
Antikörper
Type: Primary
Antigen: RSF1 (remodeling and spacing factor 1)
Clonality: monoclonal
Clone: EPR3749
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR