Anti-Tetranectin Rabbit Monoclonal Antibody [clone: EPR3067]

Lieferant: Abcam
ABCAAB108999-100EA 870 EUR
ABCAAB108999-100
Anti-Tetranectin Rabbit Monoclonal Antibody [clone: EPR3067]
Antikörper
Type: Primary
Antigen: Tetranectin
Clonality: monoclonal
Clone: EPR3067
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR