Anti-EGLN2 Rabbit monoclonal antibody unconjugated [clone: EPR2]
Lieferant: Abcam
ABCAAB108980-100EA 897 EUR
ABCAAB108980-100
Anti-EGLN2 Rabbit monoclonal antibody unconjugated [clone: EPR2]
Antikörper
Type: Primary
Antigen: EGLN2 (egl-9 family hypoxia-inducible factor 2)
Clonality: monoclonal
Clone: EPR2
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen