Anti-RAB4A Rabbit Monoclonal Antibody [clone: EPR3042]

Lieferant: Abcam
ABCAAB108974-100EA 933 EUR
ABCAAB108974-100
Anti-RAB4A Rabbit Monoclonal Antibody [clone: EPR3042]
Antikörper
Type: Primary
Antigen: RAB4A (RAB4A, member RAS oncogene family)
Clonality: monoclonal
Clone: EPR3042
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR