Anti-PRKD1 Rabbit Monoclonal Antibody [clone: EPR1492]
Lieferant: Abcam
ABCAAB108963-100EA 847 EUR
ABCAAB108963-100
Anti-PRKD1 Rabbit Monoclonal Antibody [clone: EPR1492]
Antikörper
Type: Primary
Antigen: PRKD1 (protein kinase D1)
Clonality: monoclonal
Clone: EPR1492
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen