Anti-GPA33 Rabbit Monoclonal Antibody [clone: EPR4240]

Lieferant: Abcam
ABCAAB108938-100EA 857 EUR
ABCAAB108938-100
Anti-GPA33 Rabbit Monoclonal Antibody [clone: EPR4240]
Antikörper
Type: Primary
Antigen: GPA33 (glycoprotein A33 (transmembrane))
Clonality: monoclonal
Clone: EPR4240
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR