Anti-UBE2L3 Rabbit Monoclonal Antibody [clone: EPR4368]

Lieferant: Abcam
ABCAAB108936-100EA 866 EUR
ABCAAB108936-100
Anti-UBE2L3 Rabbit Monoclonal Antibody [clone: EPR4368]
Antikörper
Type: Primary
Antigen: UBE2L3 (ubiquitin-conjugating enzyme E2L 3)
Clonality: monoclonal
Clone: EPR4368
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR