Anti-MGMT Rabbit Monoclonal Antibody [clone: EPR4397]

Lieferant: Abcam
ABCAAB108630-100EA 921 EUR
ABCAAB108630-100
Anti-MGMT Rabbit Monoclonal Antibody [clone: EPR4397]
Antikörper
Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: monoclonal
Clone: EPR4397
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR