Anti-TUBA1B Rabbit Monoclonal Antibody [clone: EPR1333]

Lieferant: Abcam
ABCAAB108629-100EA 785 EUR
ABCAAB108629-100
Anti-TUBA1B Rabbit Monoclonal Antibody [clone: EPR1333]
Antikörper
Type: Primary
Antigen: TUBA1B (tubulin, alpha 1b)
Clonality: monoclonal
Clone: EPR1333
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR