Anti-Hsp105 Rabbit Monoclonal Antibody [clone: EPR4577]

Lieferant: Abcam
ABCAAB108625-100EA 862 EUR
ABCAAB108625-100
Anti-Hsp105 Rabbit Monoclonal Antibody [clone: EPR4577]
Antikörper
Type: Primary
Antigen: Hsp105
Clonality: monoclonal
Clone: EPR4577
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR