Anti-LGALS7 Rabbit Monoclonal Antibody [clone: EPR4287]

Lieferant: Abcam
ABCAAB108623-100EA 859 EUR
ABCAAB108623-100
Anti-LGALS7 Rabbit Monoclonal Antibody [clone: EPR4287]
Antikörper
Type: Primary
Antigen: LGALS7 (lectin, galactoside-binding, soluble, 7)
Clonality: monoclonal
Clone: EPR4287
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR