Anti-ICAM3 Rabbit Monoclonal Antibody [clone: EPR3995]

Lieferant: Abcam
ABCAAB108618-100EA 878 EUR
ABCAAB108618-100
Anti-ICAM3 Rabbit Monoclonal Antibody [clone: EPR3995]
Antikörper
Type: Primary
Antigen: ICAM3 (intercellular adhesion molecule 3)
Clonality: monoclonal
Clone: EPR3995
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR