Anti-Ras Rabbit Monoclonal Antibody [clone: EPR3255]
Lieferant: Abcam
ABCAAB108602-100EA 903 EUR
ABCAAB108602-100
Anti-Ras Rabbit Monoclonal Antibody [clone: EPR3255]
Antikörper
Type: Primary
Antigen: Ras
Clonality: monoclonal
Clone: EPR3255
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen