Anti-RAD23A Rabbit Monoclonal Antibody [clone: EPR4818]

Lieferant: Abcam
ABCAAB108592-100EA 863 EUR
ABCAAB108592-100
Anti-RAD23A Rabbit Monoclonal Antibody [clone: EPR4818]
Antikörper
Type: Primary
Antigen: RAD23A (RAD23 homolog A (S. cerevisiae))
Clonality: monoclonal
Clone: EPR4818
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR