Anti-HIPK2 Rabbit Monoclonal Antibody [clone: EPR3819]
Lieferant: Abcam
ABCAAB108543-100EA 859 EUR
ABCAAB108543-100
Anti-HIPK2 Rabbit Monoclonal Antibody [clone: EPR3819]
Antikörper
Type: Primary
Antigen: HIPK2 (homeodomain interacting protein kinase 2)
Clonality: monoclonal
Clone: EPR3819
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen