Anti-IL25 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB108530-100EA 840 EUR
ABCAAB108530-100
Anti-IL25 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL25 (interleukin 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR