Anti-HSPA2 Rabbit Monoclonal Antibody [clone: EPR4596]

Lieferant: Abcam
ABCAAB108416-100EA 831 EUR
ABCAAB108416-100
Anti-HSPA2 Rabbit Monoclonal Antibody [clone: EPR4596]
Antikörper
Type: Primary
Antigen: HSPA2 (heat shock 70kDa protein 2)
Clonality: monoclonal
Clone: EPR4596
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR