Anti-CUL3 Rabbit Monoclonal Antibody [clone: EPR3195]

Lieferant: Abcam
ABCAAB108407-100EA 859 EUR
ABCAAB108407-100
Anti-CUL3 Rabbit Monoclonal Antibody [clone: EPR3195]
Antikörper
Type: Primary
Antigen: CUL3 (cullin 3)
Clonality: monoclonal
Clone: EPR3195
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR