Anti-BACE1 Rabbit Monoclonal Antibody [clone: EPR3956]
Lieferant: Abcam
ABCAAB108394-100EA 941 EUR
ABCAAB108394-100
Anti-BACE1 Rabbit Monoclonal Antibody [clone: EPR3956]
Antikörper
Type: Primary
Antigen: BACE1 (beta-site APP-cleaving enzyme 1)
Clonality: monoclonal
Clone: EPR3956
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen