Anti-ERBB2 Rabbit Monoclonal Antibody [clone: EP2324Y]

Lieferant: Abcam
ABCAAB108371-100EA 866 EUR
ABCAAB108371-100
Anti-ERBB2 Rabbit Monoclonal Antibody [clone: EP2324Y]
Antikörper
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: monoclonal
Clone: EP2324Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR