Anti-BIRC2 Rabbit Monoclonal Antibody [clone: EPR4673]

Lieferant: Abcam
ABCAAB108361-100EA 945 EUR
ABCAAB108361-100
Anti-BIRC2 Rabbit Monoclonal Antibody [clone: EPR4673]
Antikörper
Type: Primary
Antigen: BIRC2 (baculoviral IAP repeat containing 2)
Clonality: monoclonal
Clone: EPR4673
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR