Anti-ARF1 Rabbit Monoclonal Antibody [clone: EPR443]

Lieferant: Abcam
ABCAAB108347-100EA 888 EUR
ABCAAB108347-100
Anti-ARF1 Rabbit Monoclonal Antibody [clone: EPR443]
Antikörper
Type: Primary
Antigen: ARF1 (ADP-ribosylation factor 1)
Clonality: monoclonal
Clone: EPR443
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR