Anti-CASP8 Rabbit monoclonal antibody unconjugated [clone: EPR162]

Lieferant: Abcam
ABCAAB108333-100EA 939 EUR
ABCAAB108333-100
Anti-CASP8 Rabbit monoclonal antibody unconjugated [clone: EPR162]
Antikörper
Type: Primary
Antigen: CASP8 (caspase 8, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: EPR162
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR