Anti-PLIN2 Rabbit Monoclonal Antibody [clone: EPR3713]
Lieferant: Abcam
ABCAAB108323-100EA 909 EUR
ABCAAB108323-100
Anti-PLIN2 Rabbit Monoclonal Antibody [clone: EPR3713]
Antikörper
Type: Primary
Antigen: PLIN2 (perilipin 2)
Clonality: monoclonal
Clone: EPR3713
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen