Anti-CD98 Rabbit Monoclonal Antibody [clone: EPR3548]

Lieferant: Abcam
ABCAAB108300-100EA 883 EUR
ABCAAB108300-100
Anti-CD98 Rabbit Monoclonal Antibody [clone: EPR3548]
Antikörper
Type: Primary
Antigen: CD98
Clonality: monoclonal
Clone: EPR3548
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR