Anti-BRCC3 Rabbit monoclonal antibody unconjugated [clone: EPR4365]

Lieferant: Abcam
ABCAAB108295-100EA 914 EUR
ABCAAB108295-100
Anti-BRCC3 Rabbit monoclonal antibody unconjugated [clone: EPR4365]
Antikörper
Type: Primary
Antigen: BRCC3 (BRCA1/BRCA2-containing complex, subunit 3)
Clonality: monoclonal
Clone: EPR4365
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR