Anti-AGT Rabbit Monoclonal Antibody [clone: EPR2930]

Lieferant: Abcam
ABCAAB108294-100EA 833 EUR
ABCAAB108294-100
Anti-AGT Rabbit Monoclonal Antibody [clone: EPR2930]
Antikörper
Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: monoclonal
Clone: EPR2930
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR